Skip to Content
Merck
All Photos(2)

Documents

TTR009

Sigma-Aldrich

PAS TISSUE-TROL Control Slides

from human kidney

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116121
NACRES:
NA.47

biological source

human kidney

application(s)

hematology
histology

storage temp.

room temp

Application

Histology slides with paraffin embedded section of human kidney tissue. Suitable for use with Periodic Acid-Schiff (PAS) Staining System Procedure No. 395. Not tested for use in GMS procedures.

Components

Box contains 24 unstained slides plus one reference slide stained using Periodic Acid-Schiff (PAS) Staining System Procedure No. 395.

Legal Information

TISSUE-TROL is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service