SML1160
CD40-TRAF6 signaling inhibitor
≥98% (HPLC)
Synonym(s):
3-[(2,5-Dimethylphenyl)amino]-1-phenyl-2-propen-1-one, Compound 6877002
About This Item
Quality Level
Assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 20 mg/mL, clear
storage temp.
2-8°C
SMILES string
O=C(C1=CC=CC=C1)/C=N/C2=CC(C)=CC=C2C
InChI
1S/C16H15NO/c1-12-8-9-13(2)15(10-12)17-11-16(18)14-6-4-3-5-7-14/h3-11H,1-2H3/b17-11+
InChI key
WIFPLULEUNBSTE-GZTJUZNOSA-N
Application
Biochem/physiol Actions
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service