SML0981
RXFP1-Agonist-8
≥98% (HPLC)
Synonym(s):
2-Isopropoxy-N-(2-(3-(trifluoromethylsulfonyl)phenylcarbamoyl)phenyl)benzamide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C24H21F3N2O5S
CAS Number:
Molecular Weight:
506.49
UNSPSC Code:
51111800
NACRES:
NA.77
Recommended Products
Assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 10 mg/mL, clear
storage temp.
2-8°C
Biochem/physiol Actions
RXFP1-Agonist-8 is a potent selective agonist of the G-protein-coupled receptor RXFP1, human relaxin/insulin-like family peptide receptor 1,
RXFP1-Agonist-8 is a potent selective agonist of the G-protein-coupled receptor RXFP1, human relaxin/insulin-like family peptide receptor 1, with an EC50 of 94 nM in HEK293 cells and no activity with RXFP2 or RXFP3, other members of the relaxin family of receptors. RXFP1-Agonist-8 is believed to interact with an allosteric site at the ECL3 loop acting non-competitively with the natural hormone relaxin to activate RXFP1, and is the first bioavailable small-molecule agonist of human RXFP1 with good metabolic stability. Relaxin has long been known as a female hormone secreted by the corpus luteum that helps soften the cervix and relax the pelvic ligaments in childbirth. However, Relaxin also has potent cardiovascular and renal effects, inducing a 20% increase in cardiac output, 30% decrease in systemic vascular resistance, 30% increase in global arterial compliance and 45% increase in renal blood flow during pregnancy, anti-fibrotic, vasodilatory and pro-angiogenic effects that can be reproduced in both women and men with potential for treating various disorders including hypertensive diseases and acute heart failure.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service