Skip to Content
Merck
All Photos(1)

Key Documents

SML0704

Sigma-Aldrich

1-Methylnicotinamide chloride

≥98% (HPLC)

Synonym(s):

3-Carbamyl-1-methylpyridinium chloride, MNA, N-Methylnicotinic acid amide chloride, N1-Methylnicotinamide chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9ClN2O
CAS Number:
Molecular Weight:
172.61
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 15 mg/mL, clear

storage temp.

room temp

SMILES string

Cl.[n+]1(cc(ccc1)C(=N)[O-])C

InChI

1S/C7H8N2O.ClH/c1-9-4-2-3-6(5-9)7(8)10;/h2-5H,1H3,(H-,8,10);1H

InChI key

BWVDQVQUNNBTLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

1-Methylnicotinamide (MNA) is a primary metabolite of nicotinamide produced mainly by nicotinamide N-methyltransferase (NNMT). 1-Methylnicotinamide (MNA) has been shown to have antithrombotic and anti-inflammatory effects. A recent study indicated that MNA may exert antithrombotic and anti-inflammatory effects through direct action on the endothelium, with a vasorelaxing effect by enhancement of NO bioavailability and reduction of eNOS-dependent oxidative stress. Enhanced NNMT activity is a feature of many types of cancer. How much direct involvement MNA has in the tumorigenesis associated with overexpression of NNMT is under investigation. MNA by itself increased mitochondrial Complex I activity and increased cancer cell viability in SH-SY5Y neuroblastoma cells that do not produce NNMT.
1-Methylnicotinamide (MNA) is a primary metabolite of nicotinamide with antithrombotic and anti-inflammatory effects.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bashar M Thejer et al.
BMC molecular and cell biology, 21(1), 26-26 (2020-04-16)
Progesterone receptor membrane component 1 (PGRMC1) is often elevated in cancers, and exists in alternative states of phosphorylation. A motif centered on PGRMC1 Y180 was evolutionarily acquired concurrently with the embryological gastrulation organizer that orchestrates vertebrate tissue differentiation. Here, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service