Skip to Content
Merck
All Photos(1)

Key Documents

SMB00568

Sigma-Aldrich

Carviolin

≥95% (LC/MS-UV)

Synonym(s):

1,3-dihydroxy-6-(hydroxymethyl)-8-methoxy-, 9,10-Anthracenedione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H12O6
CAS Number:
Molecular Weight:
300.26
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

≥95% (LC/MS-UV)

form

solid

solubility

DMSO: 1 mg/mL

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O=C(C1=C(C=C(C=C1C2=O)O)O)C3=C2C=C(C=C3OC)CO

InChI

1S/C16H12O6/c1-22-12-3-7(6-17)2-9-14(12)16(21)13-10(15(9)20)4-8(18)5-11(13)19/h2-5,17-19H,6H2,1H3

InChI key

XNMZBRJAWRIJII-UHFFFAOYSA-N

General description

Natural product derived from fungal source.

Biochem/physiol Actions

Antibacterial activity and cytotoxicity

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeffrey E Janso et al.
Mycologia, 97(2), 444-453 (2006-01-07)
Penicillium dravuni is a new monoverticillate, sclerotium-forming species that was isolated from the alga Dictyosphaeria versluyii collected in Dravuni, Fiji. This species morphologically is similar to P. turbatum in the P. turbatum subseries of the P. thomii series of the
Tim S Bugni et al.
Journal of natural products, 67(8), 1396-1399 (2004-08-31)
Fungal isolate F01V25 was obtained from the alga Dictyosphaeria versluyii collected near Dravuni, Fiji, in 2001 and represented a previously undescribed Penicillium sp. Fermentation of isolate F01V25 resulted in the production of two new polyketides, dictyosphaeric acids A and B

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service