GE28-4082-56
HiTrap® MabSelect™
Cytiva 28-4082-56, pack of 5 × 5 mL
Synonym(s):
HiTrap MabSelect, MabSelect column
About This Item
Recommended Products
ligand
recombinant protein A (E. coli)
packaging
pack of 5 × 5 mL
manufacturer/tradename
Cytiva 28-4082-56
storage condition
(20% Ehtanol)
(20% Ethanol)
parameter
42 psi
bed size
16 mm × 25 mm
bed volume
5 mL
column I.D.
16 mm
matrix
highly cross-linked agarose
avg. part. size
85 μm (d₅₀v is the median particle size of the cumulative volume distribution)
cleaning
2-12
working range
3-10
capacity
≥30 mg binding capacity (humanlgG/ml at 2.4 min residence time)
suitability
suitable for bioprocess medium
storage temp.
2-8°C
Related Categories
General description
Application
Features and Benefits
- HiTrap® columns prepacked with MabSelect, a Bioprocess medium for capuring of Mabs from large sample volumes. • The specificity of binding to the Fc region of IgG is similar to that of native protein A, which allows one-step purification.
- Oriented coupling of recombinant Protein A to the matrix via an engineered C-terminal cysteine enhances IgG binding capacity.
- Fewer regulatory concerns due to the total absence of mammalian culture in the ligand production and purification.
Storage and Stability
Analysis Note
Other Notes
Legal Information
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Storage Class Code
3 - Flammable liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service