Skip to Content
Merck
All Photos(1)

Key Documents

AXYPDW11C

Deep well plate

size 96 wells, size 1.1 mL, pkg of 10x5plates/cs

Synonym(s):

Pipette tips for Velocity

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22

material

clear bottom
conical bottom wells

sterility

non-sterile

feature

lid: no
skirt

manufacturer/tradename

Axygen Inc P-DW-11-C
Corning

packaging

pkg of 10x5plates/cs

size

1.1 mL
96 wells

Looking for similar products? Visit Product Comparison Guide

General description

  • Axygen® deep well assay plates are ideal for use in applications including sample collection, in vitro growth chambers and long-term storage.
  • Chemically resistant and withstands temperatures down to -80°C.
  • Ultra-low profile design for reduced space requirements.
  • Flat surface for efficient sealing using heat sealing film.

Legal Information

Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service