MGVS40
Medical Viscosity Standard
viscosity 4.0 mPa.s (25.00 °C)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
Paragon Scientific Ltd
viscosity
4.0 mPa.s(25.00 °C)
application(s)
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
Find detailed technical data for all Paragon Medical Viscosity Standards on the Paragon Scientific webpage
These products contain antibacterial agent.
This product is a certified reference material (CRM) certified to International Standards BS EN ISO / IEC 17025 under UKAS accreditation
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service