BCR575
Human serum (high creatinine)
BCR®, certified reference material
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
grade
certified reference material
Agency
BCR®
manufacturer/tradename
JRC
application(s)
clinical testing
format
neat
storage temp.
−20°C
Analysis Note
For more information please see:
BCR575
BCR575
Other Notes
Sales restrictions may apply
Legal Information
BCR is a registered trademark of European Commission
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service