Skip to Content
Merck
All Photos(1)

Documents

936936

Sigma-Aldrich

SPhos Pd G4 ChemBeads

Synonym(s):

(Methanesulfonato-κO)[2′-(methylamino)-2-biphenylyl]palladium - dicyclohexyl(2′,6′-dimethoxy-2-biphenylyl)phosphine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H50NO5PPdS
CAS Number:
Molecular Weight:
794.29
MDL number:
UNSPSC Code:
31191513
NACRES:
NA.22

form

solid

Quality Level

composition

, ~5 wt. % (loading of catalyst)

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

SMILES string

CNC1=C(C2=C([Pd]OS(=O)(C)=O)C=CC=C2)C=CC=C1.COC3=CC=CC(OC)=C3C4=CC=CC=C4P(C5CCCCC5)C6CCCCC6

InChI

InChI=1S/C26H35O2P.C13H12N.CH4O3S.Pd/c1-27-23-17-11-18-24(28-2)26(23)22-16-9-10-19-25(22)29(20-12-5-3-6-13-20)21-14-7-4-8-15-21;1-14-13-10-6-5-9-12(13)11-7-3-2-4-8-11;1-5(2,3)4;/h9-11,16-21H,3-8,12-15H2,1-2H3;2-7,9-10,14H,1H3;1H3,(H,2,3,4);/q;;;+1/p-1

InChI key

ISSWOKPFLWDXSU-UHFFFAOYSA-M

General description

SPhos Pd G4 is a versatile 4th generation palladium precatalyst developed by Buchwald group.

Application

ChemBeads are chemical coated glass beads. ChemBeads offer improved flowability and chemical uniformity perfect for automated solid dispensing and high-throughput experimentation. The method of creating ChemBeads uses no other chemicals or surfactants allowing the user to accurately dispense sub-milligram amounts of chemical.

For larger scale uses, product also available in powdered form 804282

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service