Skip to Content
Merck
All Photos(6)

Key Documents

MMSMCSD48

Millipore

Milliflex Oasis® Culture Media Plate

Sabouraud dextrose media plates for yeast and mold – MXSMCSD48 to be used as replacement item. Search to explore!

Synonym(s):

Pink coded, ready-to-use agar plate for use with Milliflex Oasis® system, MILLIFLEX OASIS® Sabouraud Dextrose Media Plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NB.79

material

red polystyrene plate

Quality Level

Agency

complies with EP
complies with JP
complies with USP

sterility

sterile

form

plate (Pre-poured agar)

shelf life

9 mo.

packaging

pk of 48 tests

manufacturer/tradename

Milliflex Oasis®

pH

5.6±0.2 (25 °C)

application(s)

bioburden testing
cosmetics
pharmaceutical
water monitoring

compatibility

for use with MILLIFLEX OASIS® system

storage temp.

2-25°C

suitability

molds
yeasts

Looking for similar products? Visit Product Comparison Guide

General description

MILLIFLEX OASIS® sabouraud dextrose media plates are ready-to-use agar-based media for convenient, rapid and reliable testing. These unique pink colour coded plates offer the control of critical handling steps all along the workflow in conjugation with the MILLIFLEX OASIS® system. They are manufactured and QC tested according to pharmaceutical industry requirements. The plates can be stacked and locked to ensure safe transportation and avoid accidental opening. The empty funnel tray can also be used as an incubation rack.

MILLIFLEX® TRACE features ensures total traceability through unique 2D barcodes identification and detachable labels. If left unopened, the plates remain sealed unlike standard petri plates which reduces condensation and dehydration for an extended storage time owing to its long shelf life.

Application

MILLIFLEX OASIS® sabouraud dextrose media plates are designed for the recovery of a broad range of yeasts and molds found in various types of water, especially high purity water in the pharmaceutical industry, Beer and Bottled Water They are widely used in pharmaceutical and cosmetic industry.

Physical form

48 ready-to-use pre-poured agar plates

Analysis Note

Yeast produce white, creamy colonies. Mold are rough textured and/or filamentous. Bacteria capable of growth produce clear to white colonies

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX OASIS is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service