Skip to Content
Merck
All Photos(1)

Key Documents

MABS1982

Sigma-Aldrich

Anti-TfR1 Antibody, clone 3B82A1

clone 3B82A1, from mouse

Synonym(s):

Transferrin receptor protein 1, TR:TfR1, Trfr, T9, p90, Cd71

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
3B82A1, monoclonal
application:
FACS
ICC
IP
WB
species reactivity:
human
technique(s):
flow cytometry: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable
citations:

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

3B82A1, monoclonal

species reactivity

human

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... TFRC(7037)

Related Categories

General description

Transferrin receptor protein 1 (UniProt: P02786; also known as TR, TfR, TfR1, Trfr, T9, p90, CD71) is encoded by the TFRC gene (Gene ID: 7037) in human. Tfr is a disulfide-bonded homodimeric type II transmembrane glycoprotein that is involved in iron uptake. TfR is more abundantly expressed in rapidly dividing cells. Higher expression levels of TfR are seen in the placental trophoblast and hemoglobin-synthesizing reticulocytes, but expression is lost in mature erythrocytes. Cellular uptake of iron occurs via receptor-mediated endocytosis of ligand-occupied transferrin receptor into specialized endosomes. Endosomal acidification leads to iron release. Both TfR and TfR2 bind diferric transferrin better than apo-transferrin at neutral pH. However, TFR2 displays about 25-fold lower affinity for transferrin compared to TfR. TfR levels are regulated by cellular iron levels through binding of the iron regulatory proteins, IRP1 and IRP2, to iron-responsive elements in the 3′-UTR. Up-regulated upon mitogenic stimulation. TfR can be N- and O-glycosylated, phosphorylated and palmitoylated, however, the serum form is only glycosylated. TfR can be proteolytically cleaved on Arg100 to produce the soluble serum form (sTfR). Mutations in TFRC gene result in immunodeficiency 46, an autosomal recessive disorder that is characterized by early-onset chronic diarrhea, recurrent infections, hypo- or agammaglobulinemia, intermittent neutropenia, and thrombocytopenia.

Specificity

Clone 3B82A1 specifically detects transferrin receptor protein 1. It targets an epitope in the extracellullar domain.

Immunogen

Epitope: extracellular domain
His-tagged recombinant ectodomain of human Transferrin receptor protein 1.

Application

Detects Transferrin receptor protein 1 using this mouse monoclonal Anti-TfR1, clone 3B82A1, Cat. No. MABS1982 and has been tested for use in Flow Cytometry, Immunocytochemistry, Immunoprecipitation, and Western Blotting.
Research Category
Signaling
Western Blotting Analysis: A representative lot detected TfR1 in Western Blotting applications (Chloupkova, M., et. al. (2010). Blood Cells Mol Dis. 44(1):28-33; Johnson, M.B., et. al. (2004). Blood. 104(13):4287-93; Chen, J., et. al. (2015). J Biol Chem. 290(12):7841-50; Vogt, T.M., et. al. (2003). Blood. 101(5):2008-14).

Immunocytochemistry Analysis: A representative lot detected TfR1 in Immunocytochemistry applications (Johnson, M.B., et. al. (2007). Mol Cell Biol. 18(3):743-54; Vogt, T.M., et. al. (2003). Blood. 101(5):2008-14).

Immunoprecipitation Analysis: A representative lot detected TfR1 in Immunoprecipitation applications (Johnson, M.B., et. al. (2004). Blood. 104(13):4287-93; Vogt, T.M., et. al. (2003). Blood. 101(5):2008-14).

Quality

Evaluated by Flow Cytometry in K562 cells.

Flow Cytometry Analysis: 1 µg of this antibody detected TfR1 in 1X10E6 K562 cells.

Target description

84.87 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service