Skip to Content
Merck
All Photos(2)

Documents

FCYTMAG20KPX19BK

Millipore

MILLIPLEX® Feline Cytokine/Chemokine Magnetic Bead Panel Premixed 19 Plex, FCYTOMAG-20K-PMX Space Saver (Bulk) Packaging

Simultaneously analyze multiple cytokine and chemokine biomarkers with Bead-Based Multiplex Assays using the Luminex technology, in feline serum, plasma and cell culture samples.

Synonym(s):

feline interleukin immunoassay panel, luminex feline cytokine chemokine growth factor multiplex assay, luminex feline pro inflammatory anti inflammatory cytokine panel, millipore feline inflammation multiplex kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.47

Quality Level

species reactivity

feline

manufacturer/tradename

Milliplex®

assay range

standard curve range: 13.7-10,000 pg/mL
(sFas, IL-2, GM-CSF, IL-8, IL-12 (p40), IL-13)

standard curve range: 137.2-100,000 pg/mL
(IL-18, MCP-1)

standard curve range: 2.7-2,000 pg/mL
(KC, RANTES)

standard curve range: 274.3-200,000 pg/mL
(PDGF-BB)

standard curve range: 34.3-25,000 pg/mL
(IL-1β)

standard curve range: 6.9-5,000 pg/mL
(Flt3 Ligand)

standard curve range: 68.6-50,000 pg/mL
(IFNγ, IL-4, IL-6, SCF, SDF-1)

technique(s)

multiplexing: suitable

compatibility

configured for Premixed

detection method

fluorometric (Luminex xMAP)

shipped in

wet ice

General description

Cytokines are immunomodulatory polypeptides that play key roles in both adaptive and innate immune responses. A generic term, “cytokines” includes myokines (produced by muscle cells), lymphokines (produced by activated Th cells), interleukins (acting as mediators between T cells) and chemokines (responsible for T-cell migration). One of the regulatory mechanisms of the immune system, cytokines act at the recognition, activation, or effector phases of an immune response, modulating the development and functional activities of the subtypes of T cells, B cells and myeloid cells. Research involving cytokines plays a significant role in achieving a deeper understanding of the immune system and its multi-faceted response to most antigens, especially those responses that make up the inflammatory process.

The MILLIPLEX® Feline Cytokine/Chemokine Panel is a 19-plex premixed kit to be used for the simultaneous quantification of all the following analytes in serum, plasma, tissue/cell lysate and culture supernatant samples: Fas, Flt-3L, GM-CSF, IFNγ, IL-1β, IL-2, IL-4, IL-6, IL-8, IL-12 (p40), IL-13, IL-18, KC, MCP-1, PDGF-BB, RANTES, SCF, SDF-1, and TNFα. This kit uses a 96-well format, contains a lyophilized standard cocktail, two internal assay quality controls and can measure up to 38 samples in duplicate.

The Luminex® xMAP® platform uses a magnetic bead immunoassay format for ideal speed and sensitivity to quantitate multiple analytes simultaneously, dramatically improving productivity while conserving valuable sample volume.

Panel Type: Cytokines/Chemokines

Application

  • Analytes: sFas, Flt-3 Ligand, GM-CSF, IFN-γ, IL-1β, IL-2, IL-4, IL-6, IL-8, IL-12 (p40), IL-13, IL-18, KC, MCP-1, PDGF-BB, RANTES, SCF, SDF-1, TNF-α
  • Recommended Sample Type: Feline serum, plasma, tissue/cell lysate and culture supernatants
  • Recommended Sample
  • Dilution: 10 μL per well of undiluted serum or plasma; 25 μL tissue/cell culture samples may require dilution with an appropriate medium
  • Assay Run Time: Overnight (16-18 hours) at 2-8°C.
  • Research Category: Inflammation & Immunology

Other Notes

Sensitivity: Refer to kit protocol for sensitivities of individual biomarkers.

Standard Curve Range: Refer to QC analysis sheet for exact concentration (TNF-α)

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service