Skip to Content
Merck
All Photos(2)

Key Documents

ABN304

Sigma-Aldrich

Anti-Precerebellin Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Cerebellin precursor, Cerebellin, CER, Precerebellin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CBLN1(869)

General description

Precerebellin is the precursor of the brain-specific hexadecapeptide cerebellin, a protein with substantial similarity to the globular region of the B chain of complement component C1q. Cerebellin exerts neuromodulatory functions by directly stimulating norepinephrine release via the adenylate cyclase/pka-dependent signaling pathway; and indirectly enhances adrenocortical secretion in vivo, through a paracrine mechanism involving medullary catecholamine release. The active form of precerebellin is highly enriched in postsynaptic structures of cerebellar Purkinje cells in cartwheel neurons of the dorsal cochlear nucleus. Because precerebellin belongs to the C1q/tumor necrosis factor superfamily of secreted proteins and has similarity to adiponectin and CTRP3, it has been suggested that precerebellin possess functions other than those already stated.

Immunogen

KLH-conjugated linear peptide corresponding to human Precerebellin.

Application

Immunohistochemistry Analysis: A 1:2,000 dilution from a representative lot detected Precerebellin in human cerebellum tissue.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience
This Precerebellin antibody is validated for use in WB & IHC for the detection of the Precerebellin protein.

Quality

Evaluated by Western Blot in mouse cerebellum lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected Precerebellin in 10 µg/mL of mouse cerebellum tissue lysate.

Target description

~35 kDa observed. The calculated molecular weight is 22 kDa, however, the full length Precerebellin has been shown as a ~35 kDa band. (Bao, D., et al. (2005). Journal of Neurochemistry. 95:618-629.) An uncharacterized band may appear at ~50 kDa in some lysates.

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with 0.02% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Mouse cerebellum tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joep Beumer et al.
Cell, 181(6), 1291-1306 (2020-05-15)
Enteroendocrine cells (EECs) sense intestinal content and release hormones to regulate gastrointestinal activity, systemic metabolism, and food intake. Little is known about the molecular make-up of human EEC subtypes and the regulated secretion of individual hormones. Here, we describe an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service