Skip to Content
Merck
All Photos(1)

Key Documents

8.18492

Sigma-Aldrich

Methyl 3-oxopentanoate

for synthesis

Synonym(s):

Methyl 3-oxopentanoate, 3-Oxopentanoic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O3
CAS Number:
Molecular Weight:
130.14
MDL number:
UNSPSC Code:
12352108
EC Index Number:
250-184-3
NACRES:
NA.22

Quality Level

Assay

≥98.0% (GC)

transition temp

flash point 71 °C

density

1.04 g/cm3 at 25 °C

storage temp.

2-30°C

SMILES string

O(C)C(=O)CC(=O)CC

InChI

1S/C6H10O3/c1-3-5(7)4-6(8)9-2/h3-4H2,1-2H3

InChI key

XJMIXEAZMCTAGH-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.043 - 1.044
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service