Skip to Content
Merck
All Photos(2)

Key Documents

6015

Millipore

Amicon® Stirred Cell Manifold

The manifold is designed for use with Amicon Stirred Cells and reservoirs and allows for operation of multiple cells or reservoirs.

Synonym(s):

Maintenance Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116500
eCl@ss:
32019190

material

Delrin® plastic (chemically sanitizable)
nitrile (chemically sanitizable)

Quality Level

L × W × H

10 cm × 5 cm × 5 cm

application(s)

sample preparation

shipped in

ambient

General description

For instant direction of gas pressure or liquid flow in multi-cell or multi-reservoir systems. Can pressurize up to 3 cells or reservoirs from one gas source or feed several cells from one reservoir. Several manifolds can be combined to operate more than 3 units in parallel.

Application

With this manifold, a single feed line can be distributed to three individual outlets. Several manifolds may be grouped together to provide additional outlet streams.

Legal Information

Amicon is a registered trademark of Merck KGaA, Darmstadt, Germany
Delrin is a registered trademark of E. I. du Pont de Nemours and Company

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service