1.46208
Fraser Broth in 10ml Tubes
For enrichment of samples for Listeria sp. and Listeria monocytogene, pkg of (10 mL in 17 mL tube with blue cap (20 or 100 tubes per box))
Synonym(s):
Fraser Broth, Full Fraser Broth, ReadyTube® Fraser Broth
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Agency
ISO 11290
form
liquid (ready-to-use)
feature
ready-to-use
manufacturer/tradename
ReadyTube®
packaging
pkg of (10 mL in 17 mL tube with blue cap (20 or 100 tubes per box))
pH
7.2 ( in H2O)
application(s)
environmental
food and beverages
microbiology
storage temp.
2-8°C
suitability
Listeria spp.
Related Categories
General description
ReadyTube® is ready to use media to provide the highest level of quality and testing confidence for improving accuracy and significantly reducing the risk of false results. ReadyTube® Fraser broth is a selective enrichment medium designed to help the recovery of Listeria sp.
Application
ReadyTube® Fraser broth is used for the primary and secondary selective enrichment of Listeria sp. from food, animal feed as well as from environmental samples and other materials.
Analysis Note
This medium contains peptone, casein, yeast extract, and meat extract, which provide essential nutrients like carbon and nitrogenous compounds, including vitamins, amino acids, and trace ingredients. Phosphates buffer the medium while sodium chloride maintains osmotic equilibrium. Nalidixic acid and acriflavine inhibit the growth of Gram-negative and Gram-positive organisms respectively, except Listeria sp. Listeria sp. hydrolyze esculin to glucose and esculetin. The latter combines with ferric ions of ferric ammonium citrate, resulting in the formation of 6-7-Dihydroxycoumarin, a black-brown complex. Ferric ammonium citrate also enhances the growth of L. monocytogenes. The high salt tolerance (of sodium chloride) of Listeria is used as means to inhibit the growth of Enterococci. Lithium chloride is also used to inhibit Enterococci, which also possess the ability to hydrolyze esculin.
Legal Information
READYTUBE is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service