Skip to Content
Merck
All Photos(1)

Key Documents

1.16217

Supelco

Pre-Column holder for HPLC Columns

for LiChroCART cartridges 4-4 for Capillary connection to Hibar® RT Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115709
NACRES:
SB.52

product name

Pre-column holder for LiChroCART cartridges 4-4 for capillary connection to Hibar® RT column,

technique(s)

HPLC: suitable

Quality Level

separation technique

supercritical fluid (SFC)

storage temp.

no temp limit

General description

This precolumn holder is suitable for connection with capillary to all common HPLC columns.

Analysis Note

visual control: passes test

Legal Information

Hibar is a registered trademark of Merck KGaA, Darmstadt, Germany
LICHROCART is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service