Skip to Content
Merck
All Photos(5)

Key Documents

1.10023

Supelco

Ascorbic Acid Test

greener alternative

colorimetric, 50-2000 mg/L (ascorbic acid), for use with MQuant® StripScan App, MQuant®

Synonym(s):

Ascorbic Acid chemical test strips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

description

semi-quantitative

Quality Level

product line

MQuant®

specific analyte(s)

ascorbic acid

greener alternative product characteristics

Waste Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

measuring range

50-2000 mg/L (ascorbic acid)

compatibility

for use with MQuant® StripScan App

detection method

colorimetric

greener alternative category

storage temp.

15-25°C

General description

Method: Colorimetric, with test strips, 50 - 100 - 200 - 300 - 500 - 700 - 1000 - 2000 mg/L (ascorbic acid), MQuant®

The MQuant® Ascorbic acid test strips are suitable for quickly measuring levels of natural ascorbic acid (vitamin C) in foods such as fruit and vegetable juices, soft drinks, and also to check how much ascorbic acid has been added to certain foods as a preservative or antioxidant. Ascorbic acid reduces yellow molybdophosphoric acid to phosphomolybdenum blue. The ascorbic acid concentration is measured semi-quantitatively by visual comparison of the reaction zone of the test strip with the fields of a color scale. For a quantitative determination of Ascorbic acid please refer to our Reflectoquant® test.

Design for sustainability benefits:

We continue to demonstrate our commitment to our planet and a sustainable future by exploring ways to improve existing products in our portfolio. We have recently reviewed the current line of MQuant® test strips to identify key opportunities to minimize impact without compromising on the product performance.

This product has benefited from:

  • Test strip with reduction
  • Recycled aluminum tube
  • Digital instruction leaflet

Learn more about the initiative: Small Changes to Test Strips for Big Impact.

Linkage

Replaces: 10023-1; 10023

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service