Skip to Content
Merck
All Photos(1)

Key Documents

810127C

Avanti

18:1 NBD Lyso PE

Avanti Research - A Croda Brand 810127C

Synonym(s):

1-oleoyl-2-hydroxy-sn-glycero-3-phosphoethanolamine-N-(7-nitro-2-1,3-benzoxadiazol-4-yl) (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H50N5O10P
CAS Number:
Molecular Weight:
659.71
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810127C-500ug)

manufacturer/tradename

Avanti Research - A Croda Brand 810127C

concentration

0.5 mg/mL (810127C-500ug)

shipped in

dry ice

storage temp.

−20°C

General description

18:1 NBD Lyso PE is a fluorescent analog used as a phospholipid marker.

Packaging

5 mL Amber Glass Screw Cap Vial (810127C-500ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mitochondria: One of the origins for autophagosomal membranes?
Luo S, et al.
Mitochondrion, 9(4), 227-231 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service