Skip to Content
Merck
All Photos(1)

Key Documents

800604P

Avanti

16:0 Ethylene Glycol

1,2-dipalmitoyl ethylene glycol, powder

Synonym(s):

1,2-dihexadecanoyl ethylene glycol; DEG(16:0/16:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H66O4
CAS Number:
Molecular Weight:
538.89
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (800604P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 800604P

lipid type

neutral glycerides
neutral lipids

shipped in

dry ice

storage temp.

−20°C

InChI

1S/C34H66O4/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-33(35)37-31-32-38-34(36)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-32H2,1-2H3

InChI key

IKVCSHRLYCDSFD-UHFFFAOYSA-N

General description

16:0 Ethylene Glycol or ethylene glycol dipalmitate (EGDP) comprises two palmitic acid esterified to ethylene glycol. EGDP acts as a lubricant in plastic polymer synthesis. It is devoid of genotoxicity. Ethylene glycol is the polyols for palmitic acid ester synthesis.

Packaging

5 mL Amber Glass Screw Cap Vial (800604P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vittorio Silano et al.
EFSA journal. European Food Safety Authority, 14(10), e04580-e04580 (2016-01-01)
This statement addresses a request to EFSA from the Dutch Ministry of Public Health, Welfare and Sport to assess the impact of recent evidence underlying the conclusions of the 2016 RIVM report on the current temporary tolerable intake (t-TDI) for
Asma Chaari et al.
Journal of oleo science, 66(5), 455-461 (2017-04-07)
The production of fatty acid esters from stearic, oleic, and palmitic acids and polyols (ethylene glycol and glycerol) was investigated in this work. A series of montmorillonite-based clays catalysts (KSF, KSF/0, KP10, and K10), having different physicochemical properties, were used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service