Skip to Content
Merck
All Photos(1)

Key Documents

330841W

Avanti

MSI SPLASH

Avanti Research - A Croda Brand

Synonym(s):

MSI SPLASH LIPIDOMIX for Mass Spec Imaging

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

(MS)

form

methanol solution

packaging

pkg of 1x1 ea (330841W-1EA)

manufacturer/tradename

Avanti Research - A Croda Brand

application(s)

lipidomics

lipid type

formulation

shipped in

dry ice

storage temp.

−20°C

General description

As the role of lipids in neurodegenerative diseases such as Parkinson′s Disease is increasingly acknowledged, it has become essential to consider what lipids may have a hand in neuropathology. MSI SPLASH is made specifically for Mass Spec Imaging of neurological tissue. The mixture of thirteen internal standards is designed to be applied to brain sections using a sprayer, similarly to the application of matrix for MALDI. MSI SPLASH allows not only for quantitation of lipids within the brain, but also localization of those lipids to provide more detailed information for answering biological questions. This mixture was designed in collaboration with Michiel Vandenbosch, Shadrack Mutuku, Shane Ellis & Kim Ekroos.

Application

Mass Spectrometry

Packaging

2 mL Amber Glass Sealed Ampule (330841W-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
SPLASH is a trademark of Avanti Polar Lipids, LLC

Kit Components Also Available Separately

Product No.
Description
SDS

  • 791642C15:0-18:1-d7-PA, Avanti Research - A Croda Brand 110 μg/mLSDS

  • 791638C15:0-18:1-d7-PE, Avanti Research - A Croda Brand 100 μg/mLSDS

  • 791640C15:0-18:1-d7-PG, Avanti Research - A Croda Brand 791640C 49 μg/mLSDS

  • 791641C15:0-18:1-d7-PI, Avanti Research - A Croda Brand 23 μg/mLSDS

  • 858152L17:0-16:1 PS-d5, Avanti Research - A Croda Brand 105 μg/mLSDS

  • 856710L17:0 Lyso PE-d5, Avanti Research - A Croda Brand 3 μg/mLSDS

  • 860573PC12 Mono-Sulfo Galactosyl(β) Ceramide (d18:1/12:0), Avanti Research - A Croda Brand 860573P, powder 19 μg/mLSDS

  • 330727PC15 Lactosyl(β) Ceramide-d7 (d18:1-D7/15:0), Avanti Research - A Croda Brand 13 μg/mLSDS

  • 860677PC18 Ceramide-d7 (d18:1-d7/18:0), Avanti Research - A Croda Brand 860677P, powder 11 μg/mLSDS

  • 860569PC17 Glucosyl(β) Ceramide (d18:1/17:0), Avanti Research - A Croda Brand 860569P, powder 133 μg/mLSDS

  • 860740L18:1 SM (d18:1/18:1)-d9, Avanti Research - A Croda Brand 860740L-1MG
    860746L-1MG 31 μg/mL
    SDS

  • 855679L17:0 Lyso PC-d5, Avanti Research - A Croda Brand 3 μg/mLSDS

  • 791637C15:0-18:1-d7-PC, Avanti Research - A Croda Brand 161 μg/mLSDS

See All (13)

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service