Key Documents
Z685518
Vacuubrand chemistry pumping units, manual control, Model PC 101NT
AC/DC input 230 V AC, European 2-pin plug
Synonym(s):
evacuation pump, evaporation pump, evaporator pump
About This Item
UNSPSC Code:
41105101
NACRES:
NB.79
packaging
pack of 1 ea
manufacturer/tradename
Vacuubrand 733000
parameter
2.0-2.3 m3/h pumping speed
7 mbar vacuum
General description
This chemistry pumping unit has a wide range of applications like evacuation, evaporation and pumping of gases and vapors in chemical, biological and pharmaceutical laboratories. Typical applications are rotary evaporators, vacuum concentrators and vacuum drying ovens with high boiling solvents.
The manual flow control valve regulates the effective pumping speed at the vacuum connection; the vacuum manometer offers an analog vacuum display. The separator at the inlet (AK), made of glass with a protective coating, retains particles and liquid droplets. The exhaust waste vapor condenser enables near-100-percent solvent recovery.
  • Outstanding chemical resistance and superior vapor tolerance
  • Exceptionally high performance even at low vacuum
  • Excellent ultimate vacuum even with gas ballast
  • Manual vacuum control, analog vacuum display
  • Excellent environmental friendliness due to efficient solvent recovery
Supplied with MD 4C, manual flow control valve, analogue vacuum gauge, inlet catchpot and exhaust waste vapour condenser certification (NRTL).
Choose from one of the most recent versions:
Certificates of Analysis (COA)