Key Documents
Z268143
Sigma-Aldrich
Aldrich® desiccator cabinet
H × W × D 9 in. × 9 in. × 16 in., 2 trays, clear
About This Item
UNSPSC Code:
41104510
NACRES:
NB.43
H × W × D
9 in. × 9 in. × 16 in.
color
clear
General description
Aldrich desiccator cabinets are dust and moisture-free storage for refrigerators, freezers, and at room temperature. The clear walls, top, and base are 1/4 in. thick in acrylic for 360° visability.
Gasketed, air-tight doors are 3/8 in. thick acrylic with full length hinges and SS twist locking latches. Clear acrylic, deep slide-in trays with perforated bottoms for proper circulation within the cabinet. Dark bronze acrylic provides maximum protection for light-sensitive materials.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC