Skip to Content
Merck
All Photos(1)

Key Documents

W206008

Sigma-Aldrich

Isoamyl butyrate

≥98%, FCC, FG

Synonym(s):

3-methylbutyl butanoate, 3-methylbutyl butyrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CO2CH2CH2CH(CH3)2
CAS Number:
Molecular Weight:
158.24
FEMA Number:
2060
EC Number:
Council of Europe no.:
282
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.055
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

vapor density

5.45 (vs air)

Assay

≥98%

refractive index

n20/D 1.411 (lit.)

bp

184-185 °C (lit.)

density

0.862 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; tropical; fruity

SMILES string

CCCC(=O)OCCC(C)C

InChI

1S/C9H18O2/c1-4-5-9(10)11-7-6-8(2)3/h8H,4-7H2,1-3H3

InChI key

PQLMXFQTAMDXIZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

138.2 °F

Flash Point(C)

59 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service