SLW5100-500
Pyrex® Erlenmeyer flask, wide neck, heavy duty, with printed trace code
500 mL
Synonym(s):
Pyrex, SciLabware 5100-500
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass
flat bottom
wide-neck
Joint
joint neckWIDE-MOUTH
sterility
non-sterile
feature
graduated
without cap
heavy duty
with printed trace code
packaging
pack of 6 ea
manufacturer/tradename
SciLabware 5100-500
H
174 mm
body O.D.
97 mm
capacity
500 mL
500 mL
neck I.D.
43 mm
stopper size
43
Looking for similar products? Visit Product Comparison Guide
General description
- Mechanically stronger than conventional Erlenmeyers
- Reinforced rims for extra strength and resistance to impact fracture
- Manufactured from high quality Pyrex borosilicate glass
- White graduations and marking spot
Other Notes
Browse all Erlenmeyer flasks
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service