GF95665337
Monel® alloy 400, Ni65Cu33Fe2
foil, 50 x 50mm, 2.0mm thickness, annealed
Synonym(s):
NI110550
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
foil (annealed)
manufacturer/tradename
Goodfellow 956-653-37
size × thickness
50 mm × 50 mm × 2.0 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Legal Information
Monel is a registered trademark of Special Metals Corporation
Monel is a registered trademark of Huntington Alloys Corp.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service