GF53606875
Brass alloy, Cu63Zn37
foil, 150 x 150mm, 0.025mm thickness, hard, light tight (lt)
Synonym(s):
CU020220
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
description
light tight (lt)
form
foil (hard
hard)
manufacturer/tradename
Goodfellow 536-068-75
size × thickness
150 mm × 150 mm × 0.025 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service