Skip to Content
Merck
All Photos(1)

Key Documents

C24809

Sigma-Aldrich

3-Chlorobenzonitrile

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4CN
CAS Number:
Molecular Weight:
137.57
Beilstein:
1906694
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

99%

form

crystals

mp

38-40 °C (lit.)

SMILES string

Clc1cccc(c1)C#N

InChI

1S/C7H4ClN/c8-7-3-1-2-6(4-7)5-9/h1-4H

InChI key

WBUOVKBZJOIOAE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

206.6 °F

Flash Point(C)

97 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service