Key Documents
BR140300
BRAND® holder for separating funnel
for rods of 8 - 14 mm diam.
Synonym(s):
Accessories, Funnels, Separating funnels
About This Item
UNSPSC Code:
41121809
NACRES:
NB.35
manufacturer/tradename
BRAND 140300
Related Categories
General description
The BRAND® Holder for Separating Funnel is designed to securely support one or two funnels with a top outer diameter ranging from 50 to 120 mm. Made from durable polypropylene (PP), this holder is built to withstand the demands of laboratory environments. It can be easily mounted on support rods with an outer diameter of 8-14 mm, featuring a locking nut for added stability and security. This versatile holder enhances the efficiency of your separation processes, providing reliable support for your funnels during experiments and ensuring safe and effective liquid handling.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)