Skip to Content
Merck
All Photos(2)

Key Documents

915343

Sigma-Aldrich

Maleimide-poly(ethylene glycol)-b-poly(ε-caprolactone)

PEG average Mn 5,000, PCL average Mn 10,000

Synonym(s):

Maleimide PEG-PCL diblock copolymer, PEG-b-PCL, PEG-PCL

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C7H7N2O3[C2H4O]n[C6H10O2]mH
UNSPSC Code:
12352111
NACRES:
NA.23

form

powder or solid

Quality Level

mol wt

PCL average Mn 10,000 (by NMR)
PCL average Mn 10,000
PEG average Mn 5,000 (by NMR)
PEG average Mn 5,000

color

off-white to yellow

PDI

≤1.4 (by GPC)

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Maleimide-poly(ethylene glycol)-b-poly(ε-caprolactone) is a functionalized, amphiphilic, diblock copolymer composed of a hydrophilic PEG block and a hydrophobic PCL block. These biodegradable, biocompatible polymers can self-assemble to form nanoparticles, such as micelles and polymersomes, in both aqueous and non-aqueous media. Due to these properties, these polymers are widely used in polymeric nanoparticle formulation to achieve controlled and targeted delivery of therapeutic agents (e.g. APIs, genetic material, peptides, vaccines, and antibiotics). Additionally, well-defined nanoparticles with tunable size and properties can be prepared by altering the molecular weight ratios between hydrophilic and hydrophobic blocks, as well as by controlling formulation parameters. The carboxylic acid functional group on the PEG chain enables rapid and facile surface functionalization, allowing for these materials to be used in applications such as targeted drug delivery.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jian Jin et al.
PloS one, 9(11), e112200-e112200 (2014-11-12)
In this content, a small molecular ligand of prostate specific membrane antigen (SMLP) conjugated poly (caprolactone) (PCL)-b-poly (ethylene glycol) (PEG) copolymers with different block lengths were synthesized to construct a satisfactory drug delivery system. Four different docetaxel-loaded polymeric micelles (DTX-PMs)
Cem Varan et al.
Beilstein journal of nanotechnology, 8, 1446-1456 (2017-09-14)
Background: Brain tumors are the most common tumors among adolescents. Although some chemotherapeutics are known to be effective against brain tumors based on cell culture studies, the same effect is not observed in clinical trials. For this reason, the development

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service