Skip to Content
Merck
All Photos(2)

Key Documents

911445

Sigma-Aldrich

Polyester bis-MPA dendron, 4 Rhodamine, 1 carboxyl (core)

generation 2

Synonym(s):

Bis-MPA dendron, Fluoresenct linker, Fluoresent dendron

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C127H142Cl4N8O18
Molecular Weight:
2210.34
UNSPSC Code:
12162002
NACRES:
NA.23

form

solid

mol wt

generation 2

no. Surface Groups

4

color

red to pink

functional group

carboxylic acid (core)

storage temp.

−20°C

General description

  • Polymer architecture: Dendron
  • End Group Functionality: Rhodamine
  • Bis-MPA : 2,2-Bis(hydroxymethyl)propionic acid

Application

Bis-MPA dendritic molecules are based on 2,2-bismethylolpropionicacid (bis-MPA) building blocks. The hydrolysable linkers presented in this type of polymers allow their degradation into byproducts that can be reabsorbed by the body or into excretable non-toxic small molecules. Therefore, this group of dendritic polymers is biocompatible and biodegradable, making them suitable for biomedical applications.
The core of this product is functionalized by carboxyl groups, which can be readily used in bioconjugation. The rhodamine labeled corona allows for application in bioimaging.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patrik Stenström et al.
Molecules (Basel, Switzerland), 23(8) (2018-08-17)
Herein, we present the first evaluation of cationic dendrimers based on 2,2-bis(methylol)propionic acid (bis-MPA) as nonviral vectors for transfection of short interfering RNA (siRNA) in cell cultures. The study encompassed dendrimers of generation one to four (G1⁻G4), modified to bear
Chemistry of multifunctional polymers based on bis-MPA and their cutting-edge applications.
Garcia-Gallego S, et al
Progress in Polymer Science, 48, 85-110 (2015)
Anna Carlmark et al.
Chemical Society reviews, 42(13), 5858-5879 (2013-05-01)
Dendritic polymers are highly branched, globular architectures with multiple representations of functional groups. These nanoscale organic frameworks continue to fascinate researchers worldwide and are today under intensive investigation in application-driven research. A large number of potential application areas have been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service