Skip to Content
Merck
All Photos(1)

Key Documents

911259

Sigma-Aldrich

Poly(ethylene glycol) linear dendrimer

carboxyl terminated, PEG Mn 10k, generation 3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C588H1086O318
Molecular Weight:
13244.72
UNSPSC Code:
12352120
NACRES:
NA.23

form

solid

mol wt

PEG Mn 10k
PEG average Mn 10,000
generation 3

no. Surface Groups

16

color

white to off-white

functional group

carboxylic acid

storage temp.

−20°C

General description

  • Polymer architecture: PEG-core dendrimer
  • End Group Functionality: Carboxylic acid
  • Bis-MPA : 2,2-Bis(hydroxymethyl)propionic acid

Application

Bis-MPA dendritic molecules are based on 2,2-bismethylolpropionicacid (bis-MPA) building blocks. The hydrolyzable linker allows for degradation into byproducts that can be reabsorbed by the body or into excretable non-toxic small molecules. This dendritic polymer is biocompatible, biodegradable, and suitable for biomedical applications. PEG-core dendrimers feature excellent solubility in water and a high degree of functionality per PEG chain, enabling use for advanced drug delivery systems or for drug targeting. The carboxylic acid-functionalized corona can be readily used for bioconjugation reactions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemistry of multifunctional polymers based on bis-MPA and their cutting-edge applications
Garcia-Gallego S, et al
Progress in Polymer Science, 48, 85-110 (2015)
Anna Carlmark et al.
Chemical Society reviews, 42(13), 5858-5879 (2013-05-01)
Dendritic polymers are highly branched, globular architectures with multiple representations of functional groups. These nanoscale organic frameworks continue to fascinate researchers worldwide and are today under intensive investigation in application-driven research. A large number of potential application areas have been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service