Skip to Content
Merck
All Photos(1)

Documents

69465

Sigma-Aldrich

Dimethoxy-methyl(3,3,3-trifluoropropyl)silane

≥97.0% (GC)

Synonym(s):

Methyl(3,3,3-trifluoropropyl)dimethoxysilane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13F3O2Si
CAS Number:
Molecular Weight:
202.25
Beilstein:
1753237
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (GC)

form

liquid

refractive index

n20/D 1.358

density

1.089 g/mL at 20 °C (lit.)

SMILES string

CO[Si](C)(CCC(F)(F)F)OC

InChI

1S/C6H13F3O2Si/c1-10-12(3,11-2)5-4-6(7,8)9/h4-5H2,1-3H3

InChI key

DIJRHOZMLZRNLM-UHFFFAOYSA-N

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

136.0 °F - closed cup

Flash Point(C)

57.8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service