Skip to Content
Merck
All Photos(2)

Key Documents

677779

Sigma-Aldrich

Benzo[c][1,2,5]oxadiazole-5-boronic acid pinacol ester

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15BN2O3
CAS Number:
Molecular Weight:
246.07
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

87-91 °C

SMILES string

CC1(C)OB(OC1(C)C)c2ccc3nonc3c2

InChI

1S/C12H15BN2O3/c1-11(2)12(3,4)17-13(16-11)8-5-6-9-10(7-8)15-18-14-9/h5-7H,1-4H3

InChI key

MQXANAGMQAYTMV-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service