Skip to Content
Merck
All Photos(1)

Documents

641022

Sigma-Aldrich

2-Mercaptoethylamine, polymer-bound

70-90 mesh, extent of labeling: 0.6-1.1 mmol/g loading, 1 % cross-linked with divinylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

crosslinking

1 % cross-linked with divinylbenzene

reaction suitability

reaction type: solution phase peptide synthesis
reagent type: catalyst
reagent type: chelator

extent of labeling

0.6-1.1 mmol/g loading

particle size

70-90 mesh

storage temp.

2-8°C

Related Categories

Application

Scavenges benzyl and allyl halides and ketones.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sheppeck, J.E. et al.
Tetrahedron Letters, 41, 5329-5329 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service