Recommended Products
isotopic purity
99 atom % D
Quality Level
form
solid
mp
283-284 °C (lit.)
mass shift
M+9
SMILES string
Cl.[2H]C([2H])([2H])N(C([2H])([2H])[2H])C([2H])([2H])[2H]
InChI
1S/C3H9N.ClH/c1-4(2)3;/h1-3H3;1H/i1D3,2D3,3D3;
InChI key
SZYJELPVAFJOGJ-KYRNGWDOSA-N
Related Categories
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service