Skip to Content
Merck
All Photos(1)

Key Documents

545791

Sigma-Aldrich

2-Methyl-3-(methylthio)pyrazine

80%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8N2S
CAS Number:
Molecular Weight:
140.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

80%

refractive index

n20/D 1.585 (lit.)

bp

213-214 °C (lit.)

density

1.15 g/mL at 25 °C (lit.)

SMILES string

CSc1nccnc1C

InChI

1S/C6H8N2S/c1-5-6(9-2)8-4-3-7-5/h3-4H,1-2H3

InChI key

PPPFFGVGWFKTHX-UHFFFAOYSA-N

Other Notes

Contains 2,6 isomers, remainder 2,5 isomers

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

210.0 °F - closed cup

Flash Point(C)

98.9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silvia Maglioni et al.
Experimental gerontology, 56, 89-98 (2014-04-09)
Progressive neuronal deterioration accompanied by sensory functions decline is typically observed during aging. On the other hand, structural or functional alterations of specific sensory neurons extend lifespan in the nematode Caenorhabditis elegans. Hormesis is a phenomenon by which the body

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service