Merck
  • MiR-29b modulates DNA methylation in promoter region of miR-130b in mouse model of Diabetic nephropathy.
MiR-29b modulates DNA methylation in promoter region of miR-130b in mouse model of Diabetic nephropathy.
Journal of diabetes and metabolic disorders (2023-11-17)
Piyush Gondaliya, Kavya Jash, Akshay Srivastava, Kiran Kalia
ABSTRACT
Epigenetic modifications play a role in Diabetic Nephropathy (DN). Downregulation of miR-29b leads to modulation of DNA methylation via DNA methyl transferases (DNMTs) and hence exaggerated renal fibrosis in DN. Therefore, the main aim of the study was to evaluate effect of miR-29b expression in vivo on DNMTs, renal fibrosis, glomerular and tubular damage as well as renal morphology in DN. In order to explore the role of miR-29b in DNA methylation of other miRNAs, methylation profiling study was performed. It revealed that miR-29b was involved in methylation on of miR-130b on the cytosine guanine dinucleotides rich DNA (CpG) island 1 located on promoter region. In conclusion, miR-29b expression was found to modulate DNA methylation via DNMTs and regulate methylation of miR-130b. The result of this study provides a future direction to unveil role of miRNA expression in DNA methylation and its consequent effect on other miRNAs in DN. The online version contains supplementary material available at 10.1007/s40200-023-01208-2.
Sigma-Aldrich
Anti-DNMT3B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution