Merck
  • Insecticidal anthranilic diamides: a new class of potent ryanodine receptor activators.
Insecticidal anthranilic diamides: a new class of potent ryanodine receptor activators.
Bioorganic & medicinal chemistry letters (2005-09-17)
George P Lahm, Thomas P Selby, John H Freudenberger, Thomas M Stevenson, Brian J Myers, Gilles Seburyamo, Ben K Smith, Lindsey Flexner, Christopher E Clark, Daniel Cordova
ABSTRACT
A novel class of anthranilic diamides has been discovered with exceptional insecticidal activity on a range of Lepidoptera. These compounds have been found to exhibit their action by release of intracellular Ca2+ stores mediated by the ryanodine receptor. The discovery, synthesis, structure-activity, and biological results are presented.
Product Description
Supelco
α-Cypermethrin, PESTANAL®, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Supelco
Cypermethrin solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Supelco
Cypermethrin, PESTANAL®, analytical standard