Merck
  • Cis-selective ring-opening metathesis polymerization with ruthenium catalysts.
Cis-selective ring-opening metathesis polymerization with ruthenium catalysts.
Journal of the American Chemical Society (2012-01-14)
Benjamin K Keitz, Alexey Fedorov, Robert H Grubbs
ABSTRACT
Cis-selective ring-opening metathesis polymerization of several monocyclic alkenes as well as norbornene and oxanorbornene-type monomers using a C-H activated, ruthenium-based metathesis catalyst is reported. The cis content of the isolated polymers depended heavily on the monomer structure and temperature. A cis content as high as 96% could be obtained by lowering the temperature of the polymerization.
Sigma-Aldrich
Hoveyda-Grubbs Catalyst® M2001, Umicore