Merck
Improved ruthenium catalysts for Z-selective olefin metathesis.
Journal of the American Chemical Society (2011-11-22)
Benjamin K Keitz, Koji Endo, Paresma R Patel, Myles B Herbert, Robert H Grubbs
ABSTRACT
Several new C-H-activated ruthenium catalysts for Z-selective olefin metathesis have been synthesized. Both the carboxylate ligand and the aryl group of the N-heterocyclic carbene have been altered and the resulting catalysts evaluated using a range of metathesis reactions. Substitution of bidentate with monodentate X-type ligands led to a severe attenuation of metathesis activity and selectivity, while minor differences were observed between bidentate ligands within the same family (e.g., carboxylates). The use of nitrato-type ligands in place of carboxylates afforded a significant improvement in metathesis activity and selectivity. With these catalysts, turnover numbers approaching 1000 were possible for a variety of cross-metathesis reactions, including the synthesis of industrially relevant products.
Sigma-Aldrich
Allylboronic acid pinacol ester, 97%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hoveyda-Grubbs Catalyst® M2001, Umicore