V900042
Potassium phosphate tribasic
Vetec™, reagent grade, 97%
Synonym(s):
Tripotassium phosphate
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
K3PO4
CAS Number:
Molecular Weight:
212.27
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
Recommended Products
grade
reagent grade
product line
Vetec™
Assay
97%
density
2.564 g/mL at 25 °C (lit.)
SMILES string
[K].OP(O)(O)=O
InChI
1S/3K.H3O4P/c;;;1-5(2,3)4/h;;;(H3,1,2,3,4)/q3*+1;/p-3
InChI key
LWIHDJKSTIGBAC-UHFFFAOYSA-K
Looking for similar products? Visit Product Comparison Guide
Legal Information
Vetec is a trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Luis E Rosas-Vidal et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 39(9), 2161-2169 (2014-03-15)
Infusing brain-derived neurotrophic factor (BDNF) into the infralimbic (IL) prefrontal cortex is capable of inducing extinction. Little is known, however, about the circuits mediating BDNF effects on extinction or the extent to which extinction requires BDNF in IL. Using local
Sjef Moling et al.
The Plant cell, 26(10), 4188-4199 (2014-10-30)
Rhizobial Nod factors are the key signaling molecules in the legume-rhizobium nodule symbiosis. In this study, the role of the Nod factor receptors NOD FACTOR PERCEPTION (NFP) and LYSIN MOTIF RECEPTOR-LIKE KINASE3 (LYK3) in establishing the symbiotic interface in root
Janet E B Barber et al.
Analytical biochemistry, 463, 23-30 (2014-06-21)
D-Amino acid aminotransferase (DAAT) catalyzes the synthesis of numerous d-amino acids, making it an attractive biocatalyst for the production of enantiopure d-amino acids. To bolster its biocatalytic applicability, improved variants displaying increased activity toward non-native substrates are desired. Here, we
Anas A Al-Nabulsi et al.
Food microbiology, 46, 154-160 (2014-12-06)
Prevalence of antibiotic resistance of Listeria monocytogenes isolated from a variety of foods has increased in many countries. L. monocytogenes has many physiological adaptations that enable survival under a wide range of environmental stresses. The objective of this study was
Tomer Shpilka et al.
The EMBO journal, 34(16), 2117-2131 (2015-07-15)
Autophagy is a major catabolic process responsible for the delivery of proteins and organelles to the lysosome/vacuole for degradation. Malfunction of this pathway has been implicated in numerous pathological conditions. Different organelles have been found to contribute to the formation
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service