Y0002060
Boldine for system suitability
European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Boldine, 2,9-Dihydroxy-1,10-dimethoxyaporphine
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
boldine
manufacturer/tradename
EDQM
format
neat
storage temp.
2-8°C
SMILES string
COc1cc-2c(CC3N(C)CCc4cc(O)c(OC)c-2c34)cc1O
InChI
1S/C19H21NO4/c1-20-5-4-10-7-15(22)19(24-3)18-12-9-16(23-2)14(21)8-11(12)6-13(20)17(10)18/h7-9,13,21-22H,4-6H2,1-3H3/t13-/m0/s1
InChI key
LZJRNLRASBVRRX-ZDUSSCGKSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Packaging
Other Notes
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service