Skip to Content
Merck
All Photos(1)

Key Documents

96865

Supelco

4,4′-Dihydroxybiphenyl

analytical standard

Synonym(s):

4,4′-Biphenol, 4,4′-Biphenyldiol, 4,4′-Diphenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4C6H4OH
CAS Number:
Molecular Weight:
186.21
Beilstein:
1908886
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.21
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥95.0%

technique(s)

HPLC: suitable

mp

280-282 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

Oc1ccc(cc1)-c2ccc(O)cc2

InChI

1S/C12H10O2/c13-11-5-1-9(2-6-11)10-3-7-12(14)8-4-10/h1-8,13-14H

InChI key

VCCBEIPGXKNHFW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Self-heat. 2 - Skin Sens. 1

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 2

Flash Point(F)

537.8 °F

Flash Point(C)

281 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service