Skip to Content
Merck
All Photos(1)

Key Documents

Z706086

Sigma-Aldrich

Sigma-Aldrich® electroporation cuvettes

gap width 0.2 cm

Synonym(s):

Electroporation Cuvettes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121813
NACRES:
NB.22

capacity

~400 μL

Looking for similar products? Visit Product Comparison Guide

General description

Fits most brands of electroporators. Use for bacterial transformations, eukaryotic transfections, and the electroporation of mammalian cells and yeast. Manufactured to tight tolerances for optimum performance.
  • Use the 0.1 cm gap width for bacteria (white cap)
  • Use the 0.2 cm gap size width for yeast (blue cap)
  • Use the 0.4 cm gap size width for mammalian cells (red cap)
  • All cuvettes are individually bagged and g-irradiated for sterility.
  • The material used to manufacture the cuvettes is crystal styrene.

Legal Information

SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service