Skip to Content
Merck
All Photos(2)

Key Documents

SRP5316

Sigma-Aldrich

RIPK3, active, GST tagged human

PRECISIO®, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

RIP3

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
51111800
NACRES:
NA.32

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO®

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

30-51 nmol/min·mg

mol wt

~96 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... RIPK3(11035)

General description

Receptor interacting serine/threonine kinase 3 (RIPK3)/RIP3 is a serine/threonine kinase. It belongs to the receptor-interacting protein family. RIPK3 is characterized with a conserved amino-terminal kinase domain, which is necessary for necrosis. The gene RIPK3 is mapped to human chromosome 14. The encoded protein is composed of 518 amino acids.

Biochem/physiol Actions

Receptor interacting serine/threonine kinase 3 (RIPK3) activates necroptotic cell death and has a role in inflammatory signaling. The protein also functions in controlling viral infections, platelet functions, hemostasis and thrombosis. RIPK3 is highly expressed during cutaneous wound repair and also stimulates interleukin-1 β production.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Receptor-interacting protein kinase 3 promotes platelet activation and thrombosis.
Zhang Y, et al.
Proceedings of the National Academy of Sciences of the USA, 114(11), 2964-2969 (2017)
Receptor-interacting protein kinase 3 deficiency delays cutaneous wound healing.
Godwin A, et al.
PLoS ONE, 10(10), e0140514-e0140514 (2015)
Expression Profile and Tissue-Specific Distribution of the Receptor-Interacting Protein 3 in BALB/c Mice.
Wang Q, et al.
Biochemical Genetics, 54(4), 360-367 (2016)
RIP3: a molecular switch for necrosis and inflammation.
Moriwaki K and Chan F K M
Genes & Development, 27(15), 1640-1649 (2013)
RIPK3 restricts viral pathogenesis via cell death-independent neuroinflammation.
Daniels B P, et al.
Cell, 169(2), 301-313 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service