Skip to Content
Merck
All Photos(1)

Key Documents

SML1226

Sigma-Aldrich

YM-26734

≥95% (HPLC)

Synonym(s):

1,1′-[5-[3,4-Dihydro-7-hydroxy-2-(4-hydroxyphenyl)-2H-1-benzopyran-4-yl]-2,4,6-trihydroxy-1,3-phenylene]bis1-dodecanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H62O8
CAS Number:
Molecular Weight:
730.97
MDL number:
UNSPSC Code:
41106300
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

OC1=C(C2CC(C3=CC=C(O)C=C3)OC4=C2C=CC(O)=C4)C(O)=C(C(CCCCCCCCCCC)=O)C(O)=C1C(CCCCCCCCCCC)=O

InChI

1S/C45H62O8/c1-3-5-7-9-11-13-15-17-19-21-36(48)41-43(50)40(44(51)42(45(41)52)37(49)22-20-18-16-14-12-10-8-6-4-2)35-30-38(31-23-25-32(46)26-24-31)53-39-29-33(47)27-28-34(35)39/h23-29,35,38,46-47,50-52H,3-22,30H2,1-2H3

InChI key

CEJAYJCUSZHYDS-UHFFFAOYSA-N

Biochem/physiol Actions

YM-26734 is a competitive inhibitor of the secreted (Group II) form of phospholipase A2. The IC50 against rabbit platelet derived sPLA2 is 85 nM. YM-26734 is active against, but less potent for group I (pancreatic) sPLA2 (porcine pancreatic enzyme IC50 = 7 μM), and does not inhibit intracellular PLA2 isoforms.
YM-26734 is a natural product analog and a derivative of YM-26567-1, which is found in the fruit of Horsfieldia amygdaline. It exhibits a broad inhibitory spectrum towards different sPLA2s.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katsuhiko Hamaguchi et al.
Biochimica et biophysica acta, 1635(1), 37-47 (2003-12-04)
Although the expression of the prototypic secretory phospholipase A(2) (sPLA(2)), group IIA (sPLA(2)-IIA), is known to be up-regulated during inflammation, it remains uncertain if other sPLA(2) enzymes display similar or distinct profiles of induction under pathological conditions. In this study
Rob C Oslund et al.
Bioorganic & medicinal chemistry letters, 18(20), 5415-5419 (2008-09-27)
Simplified analogs of YM-26734, a known inhibitor of secreted phospholipase A(2) (sPLA(2)) group IIA, were synthesized and found to also display potent inhibition at low nanomolar concentrations. Analogs were based on the didodecanoylphloroglucinol portion of YM-26734 which contains the predicted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service