Skip to Content
Merck
All Photos(1)

Key Documents

SCJ4600013

Sigma-Aldrich

CF 405S succinimidyl ester

suitable for fluorescence, ≥90% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

Quality Level

Assay

≥90% (TLC)

form

solid

mol wt

~1169 g/mol (free acid)

solubility

DMF: soluble
DMSO: soluble
H2O: soluble

ε (extinction coefficient)

33000 at 404 nm

absorbance ratio

A280/404 nm 0.7

fluorescence

λex 360, 405 or 407 nm (Flow cytometry laser line)
λex 404 nm; λem 431 nm
λex 405 nm (Microscopy laser line)

suitability

suitable for fluorescence

shipped in

wet ice

storage temp.

−20°C

General description

CF 405S, succinimidyl ester (CF405S SE), a blue, fluorescent dye, is used for labeling proteins or other biomolecules having an amine group. The succinimidyl ester group of the dye reacts with an amine group to form a stable amide linkage. It has an absorption peak wavelength that nearly coincides with the 405 nm blue diode laser line. In addition, the emission peak wavelength of the dye well centers within the blue detection window of BD flow cytometers. As a result, flow cytometry analysis using CF 405S dye results in much brighter signal than that using Alexa Fluor 405.

Application

CF 405S succinimidyl ester has been used:
  • to label diphtheria toxoid (DT) to determine the percentage of DT associated with liposomes
  • in the preparation of the dye-encapsulated silica nanoparticles (CF 405S SiNP)
  • as a fluorescent dye for the fluorescent labeling of native human serum albumin (HSA) (HSAUT)

Features and Benefits

1. Much brighter than Alexa Fluor 405 due to better compatibility with both excitation and emission windows on commom instruments. 2. Highly water-soluble. 3. pH-insensitive.

Preparation Note

Protect from light and moisture

Reconstitution

Dissolve in DMSO

Legal Information

This product is distributed by Sigma-Aldrich Co. under the authorization of Biotium, Inc. This product is covered by one or more US patents and corresponding patent claims outside the US patents or pending applications owned or licensed by Biotium, Inc. including without limitation: 12/334,387; 12/607,915; 12/699,778; 12/850,578; 61/454,484. In consideration of the purchase price paid by the buyer, the buyer is hereby granted a limited, non-exclusive, non-transferable license to use only the purchased amount of the product solely for the buyer′s own internal research in a manner consistent with the accompanying product literature. Except as expressly granted herein, the sale of this product does not grant to or convey upon the buyer any license, expressly, by implication or estoppel, under any patent right or other intellectual property right of Biotium, Inc. Buyer shall not resell or transfer this product to any third party, or use the product for any commercial purposes, including without limitation, any diagnostic, therapeutic or prophylactic uses. This product is for research use only. Any other uses, including diagnostic uses, require a separate license from Biotium, Inc. For information on purchasing a license to use this product for purposes other than research, contact Biotium, Inc., 3159 Corporate Place, Hayward, CA 94545, Tel: (510) 265-1027. Fax: (510) 265-1352. Email: btinfo@biotium.com.
ALEXA FLUOR is a trademark of Life Technologies
CF is a trademark of Biotium, Inc.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tingting Chen et al.
Journal of the American Chemical Society, 135(31), 11595-11602 (2013-07-19)
A novel nanocomplex displaying single-excitation and dual-emission fluorescent properties has been developed through a crown-like assembly of dye-encapsulated silica particles decorated with satellite AuNCs for live cell imaging of highly reactive oxygen species (hROS), including •OH, ClO(-) and ONOO(-). The
Mehfuz Zaman et al.
Scientific reports, 6, 39274-39274 (2016-12-16)
The upper respiratory tract (URT) is the major entry site for human pathogens and strategies to activate this network could lead to new vaccines capable of preventing infection with many pathogens. Group A streptococcus (GAS) infections, causing rheumatic fever, rheumatic
Agnes Ulfig et al.
Redox biology, 43, 101981-101981 (2021-05-04)
Macrophages are innate immune cells that internalize and present exogenous antigens to T cells via MHC class II proteins. They operate at sites of infection in a highly inflammatory environment, generated in part by reactive oxygen species, in particular the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service