Skip to Content
Merck
All Photos(1)

Key Documents

L3019

Sigma-Aldrich

Lectin from Bandeiraea simplicifolia (Griffonia simplicifolia)

Isolectin B4 (BSI-B4), lyophilized powder

Synonym(s):

Bandeirea simplicifolia agglutinin, BS-I

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

Quality Level

form

lyophilized powder

potency

≤8 μg per mL agglutination activity (using human blood group B erythrocytes)
≥125 μg per mL agglutination activity (using human blood group A erythrocytes)

composition

Protein, ~95% E1%/280

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

BS-I has a major affinity for terminal α-D-galactosyl residues with a secondary affinity for terminal N-acetyl-α-D-galactosaminyl residues.

Other Notes

BS-I is a tetrameric lectin consisting of two types of subunits designated A and B. There are five BS-I isolectins with different subunit composition: BSI-B4, BSI-AB3, BSI-A2B2, BSI-A3B and BSI-A4. BSI-B4 is blood group B specific and has an exclusive affinity for terminal α-D-galactosyl residues, whereas BSI-A4 has blood group A specificity and has a major affinity for terminal N-acetyl-α-D-galactosaminyl residues.

Quality

Highly purified.

Physical form

Contains sodium citrate

Analysis Note

Agglutination activity is expressed in μg/mL and is determined from serial dilutions of a 1 mg/mL solution using phosphate buffered saline, pH 6.8, containing, for each lectin, calcium, magnesium, and manganese at different concentrations. This activity is the lowest concentration to agglutinate a 2% suspension of appropriate erythrocytes after 1 hr incubation at 25 °C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multiplexing Biosensor for the Detection of Extracellular Vesicles as Biomarkers of Tissue Damage and Recovery after Ischemic Stroke.
Picciolini, et al.
International Journal of Molecular Sciences, 24 (2023)
Emil Ruvinov et al.
Fibrogenesis & tissue repair, 1(1), 7-7 (2008-11-19)
Erythropoietin possesses cellular protection properties. The aim of the present study was to test the hypothesis that in situ expression of recombinant human erythropoietin (rhEPO) would improve tissue repair in rat after myocardial infarction (MI). RhEPO-producing cardiac fibroblasts were generated
Ninib Baryawno et al.
Neuro-oncology, 10(5), 661-674 (2008-08-22)
Prostaglandin E(2) (PGE(2)) has been shown to play important roles in several aspects of tumor development and progression. PGE(2) is synthesized from arachidonic acid by cyclooxygenases (COX) and prostaglandin E synthases (PGES) and mediates its biological activity through binding to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service